Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Latrunculin B, Latrunculia magnifica

Catalog No. 4280205MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-802-05MG 5 mg
42-802-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-802-05MG Supplier MilliporeSigma™ Supplier No. 4280205MG
Add to Cart
Add to Cart

A structurally unique marine toxin. Inhibits actin polymerization

A structurally unique marine toxin. Inhibits actin polymerization in vitro and disrupts microfilament organization as well as microfilament-mediated processes; 10 to 100-fold more potent than cytochalasins.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white to Yellow
Target Proteins Actin polymerization
CAS 76343-94-7
Molecular Formula C20H29NO5S
Formula Weight 395.5g/mol
Solubility DMSO, MeOH, or EtOH
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.