Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Latrunculin B, Latrunculia magnifica
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
A structurally unique marine toxin. Inhibits actin polymerization in vitro and disrupts microfilament organization as well as microfilament-mediated processes; 10 to 100-fold more potent than cytochalasins.

Specifications
Specifications
Color | Off-white to Yellow |
Target Proteins | Actin polymerization |
CAS | 76343-94-7 |
Molecular Formula | C20H29NO5S |
Formula Weight | 395.5g/mol |
Solubility | DMSO, MeOH, or EtOH |
Form | Solid |
Quantity | 1 mg |
Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction