Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Licochalcone-A, Synthetic

Catalog No. 43580010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-580-010MG 10 mg
43-580-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-580-010MG Supplier MilliporeSigma™ Supplier No. 43580010MG
Add to Cart
Add to Cart

An estrogenic flavonoid with anti-tumor and antiparasitic properties.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins Anti-tumor and antiparasitic properties
CAS 58749-22-7
Molecular Formula C21H22O4
Formula Weight 338.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Purity 96% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.