Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AICA-Riboside, 5'-Phosphate

Catalog No. 12304125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-304-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-304-125MG Supplier MilliporeSigma™ Supplier No. 12304125MG
Add to Cart
Add to Cart

A 5'-phosphorylated analog of membrane permeable AICA-Riboside that mimics AMP and acts as an activator of AMPK (AMP-activated protein kinase).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Activator of AMPK (AMP-activated protein kinase)
CAS 3031-94-5
Molecular Formula C9H15N4O8P
Formula Weight 338.2g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.