Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MEK Inhibitor I

Catalog No. 4449371MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-493-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-493-71MG Supplier MilliporeSigma™ Supplier No. 4449371MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MEK Inhibitor I, also referenced under CAS 297744-42-4, controls the biological activity of MEK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins MEK
CAS 297744-42-4
Molecular Formula C21H18N4OS
Formula Weight 374.5g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC (sum of two isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.