Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Activator II, SC79

Catalog No. 12387125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-387-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-387-125MG Supplier MilliporeSigma™ Supplier No. 12387125MG
Add to Cart
Add to Cart

Akt Activator II, SC79, CAS 305834-79-1, is a cell-permeable compound that interacts with Akt PH domain PIP3 binding pocket and makes it more susceptible to phosphorylation by upstream Kinases.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC (sum of two isomers)
Target Proteins Pleckstrin homology (PH) domain of Akt
Molecular Formula C17H17ClN2O5
Formula Weight 364.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.