Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NG-Monomethyl-L-arginine, Monoacetate Salt

Catalog No. 47588650MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-588-650MG 50 mg
47-588-625MG 25 mg
47-588-6100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 47-588-650MG Supplier MilliporeSigma™ Supplier No. 47588650MG

missing translation for 'validMainframeMsg'

Cell permeable. L-Arginine analog that acts as a competitive inhibitor of all three isoforms of NOS.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins eNOS
CAS 53308-83-1
Molecular Formula C7H16N4O2·CH3CO2H
Formula Weight 248.3g/mol
Solubility MeOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 50 mg
Purity 99% by TLC; 99% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.