Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mycophenolic Acid

Catalog No. 475913100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-591-3100MG 100 mg
47-591-3500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-591-3100MG Supplier MilliporeSigma™ Supplier No. 475913100MG
Add to Cart
Add to Cart

An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of inosine monophosphate dehydrogenase and prevents the formation of XMP and GMP.

An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of inosine monophosphate dehydrogenase and prevents the formation of XMP and GMP. Useful for selection of animal cells that express the E. coli gene coding for xanthine-guanine phosphoribosyltransferase. Also suppresses cytokine-induced nitric oxide production in mouse and rat vascular endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins Inosine monophosphate dehydrogenase
CAS 24280-93-1
Molecular Formula C17H20O6
Formula Weight 320.4g/mol
Solubility EtOH or 0.1 N NaOH
Form Solid
Quantity 100 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.