Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p-Nitroblue Tetrazolium Chloride

Catalog No. 4842351GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-423-51GM 1 g
48-423-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-423-51GM Supplier MilliporeSigma™ Supplier No. 4842351GM
Add to Cart
Add to Cart

NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4 μM).

NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4μM). A well-known scavenger of superoxide anions. Useful as a substrate for alkaline phosphatase, often in conjunction with BCIP (Cat. No. 203788).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins NOS
CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Formula Weight 817.6g/mol
Solubility MeOH or H2O
Content And Storage Hygroscopic, Protect from light
Form Crystalline Solid
Quantity 1 g
Purity 98% by TLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.