Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Okadaic Acid, Prorocentrum sp.

Catalog No. 495604100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-560-4100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-560-4100UG Supplier MilliporeSigma™ Supplier No. 495604100UG

Okadaic Acid, CAS 78111-17-8, is a highly potent inhibitor of protein phosphatase 1 (IC50 = 10-15 nM) and 2A (IC50 = 0.1 nM).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Target Proteins Protein phosphatase
CAS 78111-17-8
Molecular Formula C44H68O13
Formula Weight 805g/mol
Solubility MeOH, EtOH, DMF, CHCl3
Content And Storage Protect from light
Form Glassy Solid
Quantity 100 μg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.