Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Olomoucine

Catalog No. 4956205MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-562-05MG 5 mg
49-562-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-562-05MG Supplier MilliporeSigma™ Supplier No. 4956205MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A purine derivative that acts as a potent, reversible and selective inhibitor of p34cdk1/cyclin B and related kinases.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins p34cdk1/cyclin B
CAS 101622-51-9
Molecular Formula C15H18N6O
Formula Weight 298.3g/mol
Solubility DMSO
Form Crystalline Solid
Quantity 5 mg
Purity 98% by TLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.