Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Inhibitor XII, Isozyme-Selective, Akti-2

Catalog No. 1240292MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-402-92MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-402-92MG Supplier MilliporeSigma™ Supplier No. 1240292MG
Add to Cart
Add to Cart

The Akt Inhibitor XII, Isozyme-Selective, Akti-2 controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC (mixture of regioisomers)
Molecular Formula C39H39N7O2·3HCl
Formula Weight 747.2g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.