Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 4α-Phorbol-12,13-didecanoate

Catalog No. 5243945MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-439-45MG 5 mg
52-439-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-439-45MG Supplier MilliporeSigma™ Supplier No. 5243945MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inactive on mouse skin as an inflammatory and tumor-promoting agent.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins A negative control for Phorbol-12,13-didecanoate (PDD) and Phorbol-12-myristate-13-acetate (PMA)
CAS 27536-56-7
Molecular Formula C40H64O8
Formula Weight 672.9g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Purity >99% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.