Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Inhibitor XVI, AT7867

Catalog No. 12403310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-403-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-403-310MG Supplier MilliporeSigma™ Supplier No. 12403310MG
Add to Cart
Add to Cart

A cell-permeable pyrazolo compound that acts as a potent inhibitor against Akt (IC50 = 32, 17, and 47nM against Akt1, Akt2, and Akt3, respectively), PKA (IC50 = 20nM), and p70S6K (IC50 = 85nM) in an ATP-competitive manner (Ki = 18nM using AKT2), with much reduced potency toward RSK1 (IC50 >100nM) and a panel of 13 other cellular kinases

A cell-permeable pyrazolo compound that acts as a potent inhibitor against Akt (IC50 = 32, 17, and 47nM against Akt1, Akt2, and Akt3, respectively), PKA (IC50 = 20nM), and p70S6K (IC50 = 85nM) in an ATP-competitive manner (Ki = 18nM using AKT2), with much reduced potency toward RSK1 (IC50 >100nM) and a panel of 13 other cellular kinases (IC50 >1μM). Shown to reduce cellular phosphorylation of GSK-3β (IC50 range from 2.08 to 4.45μM), S6R, FKHRL1, and inhibit the proliferation of cancer cells exhibiting constitutive Akt pathway activation both in vitro (IC50 range from 0.94 to 11.86μM) and in mice in vivo (20mg/kg via i.p. or 90mg/kg via p.o.).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Molecular Formula C20H20ClN3
Formula Weight 337.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.