Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Albumin, Human Serum, 30% Aqueous Solution
GSA_VA

Catalog No. 1266750ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-667-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-667-50ML Supplier MilliporeSigma™ Supplier No. 1266750ML
Only null left
Add to Cart
Add to Cart

Suitable for use as Protein base or filler Antigen

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Standard, Protein Base or Filler, Blocking Agent
Formulation 30% solution in 20mM Tris-HCl, 0.1% NaN3, pH 6-8
Quantity 50 mL
Source Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
Storage Requirements 2° to 8°C
Form Liquid
Purity or Quality Grade ≥95% by agarose electrophoresis

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.