Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ALDH3A1 Inhibitor, CB29

Catalog No. 5311700001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-117-00001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-117-00001 Supplier MilliporeSigma™ Supplier No. 5311700001
Add to Cart
Add to Cart

A cell permeable, selective, reversible, substrate-binding site directed inhibitor of aldehyde dehydrogenase ALDH3A1 (Ki = 4.7 μM, IC50 = 16 μM).

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins ALDH3A1
Molecular Formula C15H15N3O5S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.