Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ALLM

Catalog No. 20872125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-872-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-872-125MG Supplier MilliporeSigma™ Supplier No. 20872125MG
Add to Cart
Add to Cart

Cell permeable inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 600 pM).

  • Peptide Sequence: N-Acetyl-Leu-Leu-Met
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description >95% by HPLC
Target Proteins calpain-1
CAS 110115-07-6
Molecular Formula C19H35N3O4S
Formula Weight 401.6g/mol
Solubility DMSO, MeOH or EtOH
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.