Learn More
MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set V, Animal-Free
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Product Type | Protease Inhibitor Cocktail |
| Target Proteins | Serine and cysteine proteases, but not metalloproteases |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Lyophilized |
| Quantity | 1 Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.