Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PP2

Catalog No. 5295735MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-957-35MG 5 mg
52-957-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-957-35MG Supplier MilliporeSigma™ Supplier No. 5295735MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, & 100 nM for p56lck, p59fynT, Hck, & Src, respectively).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Product Type Inhibitor
Target Proteins p56lck
CAS 172889-27-9
Molecular Formula C15H16ClN5
Formula Weight 301.8g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.