Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set I

Catalog No. 5391311VL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Vial
10 Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-913-11VL 1 Vial
53-913-110VL 10 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-913-11VL Supplier MilliporeSigma™ Supplier No. 5391311VL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.

A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.