Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Amiloride, Hydrochloride

Catalog No. 129876100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-987-6100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-987-6100MG Supplier MilliporeSigma™ Supplier No. 129876100MG
Add to Cart
Add to Cart

An inhibitor of angiogenesis.

An inhibitor of angiogenesis. Inhibits capillary morphogenesis completely and reversibly at approximately 130μM. At low concentrations, acts as a potent and specific inhibitor of transmembrane Na+ entry and Na+,K+-ATPase. Also acts as an inhibitor of urokinase-type plasminogen activator (uPA). Blocks smooth muscle responses to contractile stimuli and palytoxin-induced conductance in skeletal muscle. Reduces electrical potential across tubular epithelium. At higher concentrations, blocks the Na+/H+ exchange pathway.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white to Yellow
Description 98% by HPLC
Target Proteins capillary morphogenesis
CAS 2016-88-8
Molecular Formula C6H8ClN7O·HCl
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.