Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 328437

Catalog No. 5594065MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-940-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-940-65MG Supplier MilliporeSigma™ Supplier No. 5594065MG

A highly potent and selective CC chemokine receptor-3 (CCR3) antagonist.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins CC chemokine receptor-3 (CCR3)
CAS 247580-43-4
Molecular Formula C21H18N2O5
Formula Weight 378.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.