Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ γ-Secretase Inhibitor IX

Catalog No. p-7060160 Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
10 mg

InSolution™ g-Secretase Inhibitor IX, CAS 208255-80-5, is a 25 mM (5 mg/462 μl or 10 mg/924 μl) solution of g-secretase Inhibitor IX (DAPT; Cat. No. 565770) in DMSO.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Formulation A 25 mM (5mg/462μl or 10mg/924μl) solution of γ-Secretase Inhibitor IX in DMSO.
Target Proteins Aβtotal
Molecular Formula C23H26F2N2O4
Formula Weight 432.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.