Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Serine Protease Inhibitor Cocktail Set I

Catalog No. 5650005VL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Vial
5 Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-500-05VL 5 Vials
56-500-01VL 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-500-05VL Supplier MilliporeSigma™ Supplier No. 5650005VL
Add to Cart
Add to Cart

The Serine Protease Inhibitor Cocktail Set I controls the biological activity of Serine Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cocktail of four protease inhibitors that is useful for inhibition of a broad range of serine proteases. When reconstituted with 1mL H2O each vial will contain a 100X stock solution of protease inhibitors. When diluted to 1X the protease inhibitor cocktail will contain 500μM AEBSF, HCl (Cat. No. 101500), 420nM Aprotinin (Cat. No. 616370), 20μM Elastinal (Cat. No. 324691), and 1μM GGACK (Cat. No. 347436). Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins broad range of serine proteases
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 5 Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.