Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor IX

Catalog No. 5657705MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-577-05MG 5 mg
56-577-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-577-05MG Supplier MilliporeSigma™ Supplier No. 5657705MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC50 = 115 nM, Aβ42 IC50 = 200 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Product Type Inhibitor
Target Proteins Aβtotal
CAS 208255-80-5
Molecular Formula C23H26F2N2O4
Formula Weight 432.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 5 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.