Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AMPK Signaling Activator XIII, ZLN005

Catalog No. 5043770001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-437-70001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-437-70001 Supplier MilliporeSigma™ Supplier No. 5043770001
Add to Cart
Add to Cart

Calbiochem AMPK Activator, CAS 4529-96-8, inhibits luciferase activity, and activates AMPK signaling.

A cell-permeable benzimidazole that, in addition to its inhibitory potency against luciferase activity (IC50 <10nM), also activates AMPK signaling by increasing cellular ADP/ATP ratio via mitochondria respiration upcoupling, effectively activating AMPK & PGC-1α in muscle, but not liver, cell types both in cultures (5-20μM in rat L6 myotubes) in vitro and in db/db mice (15mg/kg/day via p.o.) in vivo . Reported to improve glucose and insulin tolerance of db/db mice without any apparent effects on body weight or food intake.
TRUSTED_SUSTAINABILITY

Specifications

Color Pinkish-white
Description 99% by HPLC
Target Proteins increases cellular ADP/ATP ratio via mitochondria respiration uncoupling
Molecular Formula C17H18N2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.