Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Amyloid Oligomer Inhibitor, K01-162

Catalog No. 20048710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-048-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-048-710MG Supplier MilliporeSigma™ Supplier No. 20048710MG
Add to Cart
Add to Cart

The β-Amyloid Oligomer Inhibitor, K01-162 controls the biological activity of β-Amyloid Oligomer.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 95% by HPLC
Molecular Formula C15H14BrN
Formula Weight 288.18g/mol
Solubility DMSO
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Syrup
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.