Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ S1P1 Receptor Agonist, SEW2871

Catalog No. 5677331MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-773-31MG 1 mg
56-773-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-773-31MG Supplier MilliporeSigma™ Supplier No. 5677331MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

The S1P1 Receptor Agonist, SEW2871, controls the biological activity of S1P1 Receptor. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins S1P1 Receptor
CAS 256414-75-2
Molecular Formula C20H10F6N2OS
Formula Weight 440.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.