Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Amyloid Precursor Protein β-Secretase Inhibitor

Catalog No. 171601500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-160-1500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-160-1500UG Supplier MilliporeSigma™ Supplier No. 171601500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Amyloid Precursor Protein β-Secretase Inhibitor controls the biological activity of Amyloid Precursor Protein β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Peptide Sequence: H-Lys-Thr-Glu-Glu-Ile-Ser-Glu-Val-Asn-Stat-Val-Ala-Glu-Phe-OH [Stat = (3S,4S)-Statine]
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Product Type Inhibitor
Target Proteins APP
Molecular Formula C73H118N16O27
Formula Weight 1651.8g/mol
Solubility Aqueous Buffers, pH 7.5
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.