Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Staurosporine, Streptomyces sp.

Catalog No. 569397250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-939-7250UG 250 μg
56-939-7100UG 100 μg
56-939-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 56-939-7250UG Supplier MilliporeSigma™ Supplier No. 569397250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC50 = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White to Pale Yellow
Product Type Inhibitor
Target Proteins PKA
CAS 62996-74-1
Molecular Formula C28H26N4O3
Formula Weight 466.5g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 250 μg
Purity 99% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.