Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Anaphase-Promoting Complex Inhibitor Negative Control, AAME

Catalog No. 17210350MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-210-350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-210-350MG Supplier MilliporeSigma™ Supplier No. 17210350MG
Add to Cart
Add to Cart

The Anaphase-Promoting Complex Inhibitor Negative Control, AAME, also referenced under CAS 1784-05-0, controls the biological activity of Anaphase-Promoting Complex.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by TLC
CAS 1784-05-0
Molecular Formula C9H18N4O3·HCl
Formula Weight 266.7g/mol
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.