Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor VI, S3I-201

Catalog No. 57310210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-310-210MG 10 mg
57-310-225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57-310-210MG Supplier MilliporeSigma™ Supplier No. 57310210MG
Add to Cart
Add to Cart

STAT3 Inhibitor VI, S3I-201, CAS 501919-59-1, is a cell-permeable compound that binds Stat3-SH2 domain and blocks Stat3 phosphorylation, dimerization, DNA-binding, & Stat3-dependent transcription.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
CAS 501919-59-1
Molecular Formula C16H15NO7S
Formula Weight 365.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.