Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor VI, S3I-201

Catalog No. p-7060169 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
25 mg

STAT3 Inhibitor VI, S3I-201, CAS 501919-59-1, is a cell-permeable compound that binds Stat3-SH2 domain and blocks Stat3 phosphorylation, dimerization, DNA-binding, & Stat3-dependent transcription.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
CAS 501919-59-1
Molecular Formula C16H15NO7S
Formula Weight 365.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.