Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Andrographolide

Catalog No. 17206050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-206-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-206-050MG Supplier MilliporeSigma™ Supplier No. 17206050MG
Add to Cart
Add to Cart

A bicyclic diterpenoid lactone that displays anti-viral, anti-inflammatory, anti-apoptotic, and anti-hyperglycemic properties.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by TLC
Target Proteins NF-κB and AP-1 activation
CAS 5508-58-7
Molecular Formula C20H30O5
Formula Weight 350.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.