Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Anthrax Lethal Factor Protease Inhibitor, In-2-LF

Catalog No. 1769011MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-690-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-690-11MG Supplier MilliporeSigma™ Supplier No. 1769011MG
Only null left
Add to Cart
Add to Cart

The Anthrax Lethal Factor Protease Inhibitor, In-2-LF controls the biological activity of Anthrax Lethal Factor Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Gly-Tyr-βAla-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Val-Leu-Arg-NHOH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins anthrax LF metalloprotease
Molecular Formula C81H150N42O17
Formula Weight 1984.4g/mol
Solubility DMSO, 5% Acetic acid, or H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.