Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Paclitaxel, Taxus sp.

Catalog No. 5805555MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-055-55MG 5 mg
58-055-525MG 25 mg
58-055-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58-055-55MG Supplier MilliporeSigma™ Supplier No. 5805555MG
Add to Cart
Add to Cart

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Antitumor and antileukemic agent isolated from the bark of the yew tree.

Antitumor and antileukemic agent isolated from the bark of the yew tree. Promotes assembly of microtubules and inhibits tubulin disassembly. An effective agent in blocking HeLa and B16 cells at the G2/M stage. Inhibits DNA synthesis and stimulates the release of tumor necrosis factor-α. Induces apoptosis in murine mammary carcinoma MCA-4 and ovarian carcinoma OCA-1 tumors.
TRUSTED_SUSTAINABILITY

Spécifications

Color White
Product Type Inhibitor
Target Proteins Promotes assembly of microtubules and inhibits tubulins disassembly
CAS 33069-62-4
Molecular Formula C47H51NO14
Formula Weight 853.9g/mol
Solubility DMSO or MeOH
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 5 mg
Purity 97% by HPLC
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.