Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Anthrax Lethal Factor, Recombinant, Bacillus anthracis, B. anthracis

Catalog No. 176900100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-690-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-690-0100UG Supplier MilliporeSigma™ Supplier No. 176900100UG

Recombinant, Bacillus anthracis anthrax lethal factor expressed in a specialized strain of B.

Recombinant, Bacillus anthracis anthrax lethal factor expressed in a specialized strain of B. anthracis . One of the three protein components of Anthrax toxin, lethal factor (LF) is a highly specific protease that cleaves members of the mitogen-activated protein kinase kinase (MAPKK) family. LF is comprised of four domains. Domain I binds the protective antigen to enter the target cell; domains II, III, and IV create a long groove to hold and cleave the MAPKK proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 50 mM NaCl, 5 mM HEPES, pH 7.5.
Description Single major band by SDS-PAGE
Target Proteins MAPKK
Formula Weight 90000g/mol
Solubility Sterile dH2O
Form Lyophilized
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.