Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Antipain, Dihydrochloride

Catalog No. 17822310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-822-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-822-310MG Supplier MilliporeSigma™ Supplier No. 17822310MG
Add to Cart
Add to Cart

Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes.

Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes. Inhibitor of Ca2+-dependent endopeptidases. Has specificity similar to Leupeptin (Cat. No. 108975). Inhibits trypsin-like serine proteases, papain and some cysteine proteases (IC50 = 300μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins trypsin-like serine proteases, papain, and some cysteine proteases
CAS 37682-72-7
Molecular Formula C27H44N10O6·2HCl
Formula Weight 677.6g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.