Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor I

Catalog No. 6276105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-761-05MG 5 mg
62-761-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 62-761-05MG Supplier MilliporeSigma™ Supplier No. 6276105MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-VAD-FMK, CAS 187389-52-2, is a cell-permeable, irreversible, pan-caspase inhibitor.

  • Peptide Sequence: Z-Val-Ala-Asp(OMe)-CH2F*
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Fas-mediated apoptosis in Jurkat cells and staurosporine-induced cell death in corneal epithelial cells
CAS 187389-52-2
Molecular Formula C22H30FN3O7
Formula Weight 467.5g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.