Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 203580, Hydrochloride

Catalog No. 5593951MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-939-51MG 1 mg
55-939-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-939-51MG Supplier MilliporeSigma™ Supplier No. 5593951MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB 203580, Hydrochloride, CAS 869185-85-3, is a water-soluble form of SB 203580. A highly potent, selective, ATP-competitive inhibitor of p38 MAP Kinase (IC50 = 34nM in vitro, 600nM in cells).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
Target Proteins p38 MAP kinase
Molecular Formula C21H16FN3OS·HCl
Formula Weight 413.9g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.