Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Antithrombin III, Human Plasma

Catalog No. 1697561MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-975-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-975-61MG Supplier MilliporeSigma™ Supplier No. 1697561MG
Add to Cart
Add to Cart

Single-chain glycoprotein present in normal serum at about 23mg/100 ml.

Single-chain glycoprotein present in normal serum at about 23mg/100 ml. Principal physiological inhibitor of thrombin. Present at higher levels in plasma than in serum because it complexes with thrombin during coagulation. Functions by inhibiting proteolytic enzymes involved in blood coagulation and fibrinolysis, including Factor Xa, plasmin, thrombin, and trypsin. Potency is strongly enhanced by heparin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100 mM sodium citrate buffer, 20 mM Tris-HCl, pH 8.3.
Description single band by SDS-PAGE
Target Proteins thrombin
CAS 90170-80-2
Formula Weight 58000g/mol
Solubility H2O
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.