Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AP-1/NF-κB Dual Inhibitor, SP100030

Catalog No. 5315350001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-153-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-153-50001 Supplier MilliporeSigma™ Supplier No. 5315350001
Add to Cart
Add to Cart

A cell-permeable, non-toxic, conformationally restricted pyrimidinecarboxamide compound that acts as a potent, reversible T-cell specific inhibitor of both AP-1 and NF-κB transcriptional activation (IC50 = 50

A cell-permeable, non-toxic, conformationally restricted pyrimidinecarboxamide compound that acts as a potent, reversible T-cell specific inhibitor of both AP-1 and NF-κB transcriptional activation (IC50 = 50nM). Reduces DNA binding activity of NF-κB and downregulates NF-κB driven cytokine gene expression. Displays only trivial inhibitory effect against cytokine induction in a variety of other cells. Exhibits anti-inflammatory activity in a murine model of collagen-induced arthritis (∽ 10mg/kg/d, i.p.) and suppresses the expansion of AH-130 Yoshida ascites hepatoma in rats (∽ 5mg/kg/d, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins AP-1 and NF-κB
Molecular Formula C14H5ClF9N3O
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.