Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Withaferin A, Withania somnifera

Catalog No. 6815355MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-153-55MG 5 mg
68-153-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-153-55MG Supplier MilliporeSigma™ Supplier No. 6815355MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable steroidal lactone that displays antitumor, anti-inflammatory, radiosensitizing, and immunosuppressive properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins A steroidal lactone that displays antitumor, anti-inflammatory, radiosensitizing, and immunosuppressive properties.
CAS 5119-48-2
Molecular Formula C28H38O6
Formula Weight 470.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 94% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.