Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Zebularine

Catalog No. 69140025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-140-025MG 25 mg
69-140-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-140-025MG Supplier MilliporeSigma™ Supplier No. 69140025MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable chemically stable cytidine analog that displays antitumor properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Product Type Inhibitor
Target Proteins Cytidine deaminase
CAS 3690-10-6
Molecular Formula C9H12N2O5
Formula Weight 228.2g/mol
Solubility DMSO or MeOH
Form Solid
Quantity 25 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.