Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Apigenin

Catalog No. 1782785MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-827-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-827-85MG Supplier MilliporeSigma™ Supplier No. 1782785MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Induces the reversion of transformed phenotypes of v-H-ras-transformed NIH 3T3 cells at low concentration (12.5 μM).

Induces the reversion of transformed phenotypes of v-H- ras -transformed NIH 3T3 cells at low concentration (12.5μM). This finding has been attributed to the inhibition of MAP kinase activity. Inhibits the proliferation of malignant tumor cells by G2/M arrest and induces morphological differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins MAP kinase
CAS 520-36-5
Molecular Formula C15H10O5
Formula Weight 270.2g/mol
Solubility hot alcohol or DMSO
Form Crystalline Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.