Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ApoE4 Modulator, PH002

Catalog No. 17847710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-847-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-847-710MG Supplier MilliporeSigma™ Supplier No. 17847710MG
Add to Cart
Add to Cart

A phthalazinone derived compound that directly binds to the 22-kDa amino-terminal region of apolipoprotein E4 (ApoE4) and blocks its intramolecular domain interactions in neuronal cells thereby reversing the detrimental effects of ApoE4.

A phthalazinone derived compound that directly binds to the 22-kDa amino-terminal region of apolipoprotein E4 (ApoE4) and blocks its intramolecular domain interactions in neuronal cells (IC50 = 116nM) thereby reversing the detrimental effects of ApoE4. Established to be more potent than GIND25. Restores mitochondrial functions and elevates the levels of complex IV subunit 1. Shown to increase in COX1 levels in primary neurons from NSE-ApoE4 transgenic mouse cortex and hippocampus. Shown to rescue impairments of mitochondrial motility in primary neurons from NSE-ApoE4 transgenic mice (∽ 200nM). Also restores endoplasmic reticulum and Golgi apparatus transit of ApoE4 in cultured neurons and brings it to levels normally found in ApoE3 and ApoE4-R61T.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins APOE4
Formula Weight 491.58g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.