Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Apoptosis Activator III, Embelin

Catalog No. 17849310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-849-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-849-310MG Supplier MilliporeSigma™ Supplier No. 17849310MG
Add to Cart
Add to Cart

The Apoptosis Activator III, Embelin, also referenced under CAS 550-24-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description Single spot by TLC
Target Proteins BIR3
CAS 550-24-3
Molecular Formula C17H26O4
Formula Weight 294.4g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.