Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Apoptosis Activator VI, CD437/AHPN

Catalog No. 1784965MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-849-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-849-65MG Supplier MilliporeSigma™ Supplier No. 1784965MG
Add to Cart
Add to Cart

The Apoptosis Activator VI, CD437/AHPN, also referenced under CAS 125316-60-1, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
CAS 125316-60-1
Molecular Formula C27H26O3
Formula Weight 398.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.