Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ APT1 Inhibitor II, Cpd21

Catalog No. 5316200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-162-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-162-00001 Supplier MilliporeSigma™ Supplier No. 5316200001
Add to Cart
Add to Cart

A cell permeable acetylpiperazine derivative compound that acts as a selective and reversible inhibitor of lysophospholipase 1 (LYPLA1; IC50 = 840 nM, Ki = 300nM) without affecting the activity of LYPLA2 and 25 other serine hydrolases in mouse BW5147 T cell hydridoma pr

A cell permeable acetylpiperazine derivative compound that acts as a selective and reversible inhibitor of lysophospholipase 1 (LYPLA1; IC50 = 840 nM, Ki = 300nM) without affecting the activity of LYPLA2 and 25 other serine hydrolases in mouse BW5147 T cell hydridoma proteome. Shown to cause an almost complete inhibition (+90%) of LYPLA1 activity in lung, heart, and kidney (50mg/kg, 3 h), and in HEK293T, and mouse T cells (∽ 5μM for 3 h).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins APT1
Molecular Formula C18H17ClF3N3O3
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.