Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Aromatase Inhibitor I

Catalog No. 1825401MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-254-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-254-01MG Supplier MilliporeSigma™ Supplier No. 1825401MG
Add to Cart
Add to Cart

The Aromatase Inhibitor I, also referenced under CAS 331684-05-0, controls the biological activity of Aromatase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins P450 arom.
CAS 331684-05-0
Molecular Formula C17H11N3O4
Formula Weight 321.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.